Would you like to log in again?
Cardionics® E-Scope Electronic Stethoscope
Supplier: AMERICAN 3B SCIENTIFIC SE
1022457 1022487 1022486 1022466 1022458 1022459 1022489 1022456 1022488 1022397 1022465 1021985
470318-020EA 427.29 USD InStock
470318-020 470318-118 470318-108 470318-114 470318-104 470318-116 470318-106 470318-110 470318-100 470318-112 470318-102 470318-036
Cardionics® E-Scope Electronic Stethoscope
Stethoscopes
Magnify Auscultatory sounds.
  • Can be used with hearing aid or hearing impairment
  • One-handed volume control
  • Specialist adult diaphragm
  • Automatic shut off
  • Audio output jack
The E-Scope (Electronic Stethoscope) Clinical Model allows medical professionals to auscultate with increased volume by amplifying sounds up to 30 times louder than an acoustic scope. The E-Scope is popular around the world in clinics, hospitals, research laboratories, and telemedicine settings. Additionally, the E-Scope can be paired with an accessory cord to broadcast sounds through speakers or other audio outputs.
The Standard E-Scope ships with traditional binaural. Additional headphone, or wired connections, can provide accommodation for individuals with hearing impairments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty