Would you like to log in again?
3B Scientific® Image Guided Cervical Injection Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1021900
470318-132EA 6754.39 USD InStock
470318-132
3B Scientific® Image Guided Cervical Injection Trainer
Models Human Biology Models
An economic alternative to cadaver practice.
  • Life-Like radiopacity for Realistic X-Ray Images
  • Realistic injection haptics
  • Anatomically accurate bone structure
  • Visually identifiable landmarks
The Image Guided Cervical Spinal Injection Trainer enables trainees to develop a three-dimensional understanding of the procedures for successful spine interventions. They will learn to correlate imaging with the cervical spinal anatomy relevant to interventional pain procedures, to identify the target tissue and vulnerable structures using imaging and anatomic inspection, and to apply best practices to ensure patient safety during the interventional spine procedures.
Accessories information: See additional information for feature details.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty