Would you like to log in again?
Rainbow Electrolysis, Aldon
Supplier: Aldon
IS7046
470320-334KT 19.4 USD InStock
470320-334
Rainbow Electrolysis, Aldon
Educational Classroom Kits and Activities
Aligned to the next generation science standards.
  • Disciplinary Core Ideas: PS1.A; PS1.B
  • Performance Expectations: MS-PS1-2; HS-PS1-2
  • Cross Cutting Concepts: Patterns
Engineering practices: Analyzing and interpreting data; scientific knowledge is based on empirical evidence; constructing explanations and designing solutions.
Study oxidation-reduction, electrolysis and acid-base reactions as Universal Indicator is added to an aqueous sodium sulfate solution. When two carbon electrodes are connected to a battery and placed in the solution, a rainbow of colors forms as the water is electrolyzed. There are enough materials to complete the demonstration five times.
Delivery information: Kit includes: 50 ml Sodium Sulfate Solution, 1 m, 6 ml Universal Indicator, 1 red connecting wire, 1 black connecting wire, 12 carbon electrodes and 2 petri dishes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty