Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Amino Labs BioInk Bacterial Paint Kits
Supplier: AMINO LABS NORTH INCORPORATED SE
WWA011-S WWA011-G_CA WWA011-S_CA WWA011-G
470320-390EA 496.09 USD InStock
CA470318-708 470320-390 CA470318-710 470320-388
Amino Labs BioInk Bacterial Paint Kits
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Create prints and textile art with colorful bacteria.
  • Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
Grow patterns, paintings and symbols on petri dishes using the colored bacteria provided in your kit (as well as any colored engineered bacteria you may have created before). Once grown, transfer them directly to paper or textile in any design you like, or use some of the paintbrushes to add some freehand elements to your art. Finish by stabilizing and fixing your creation(s) with the medium provided to lock in the colorful creation.
Keep your engineered-bacteria immortalized with this Keep-it Kit™ (available separately). You can even touch it once it sets!
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty