Would you like to log in again?
Amino Labs Canvas Kit™
Supplier: AMINO LABS NORTH INCORPORATED SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WWA003-G WWA003-S_CA WWG012-S_CA WWA003-G_CA WWA003-GR WWG012-G WWA003-GR_CA WWG012-G_CA WWA003-SR WWA003-S WWA003-SR_CA WWG012-S
470320-392EA 23 USD InStock
470320-392 470320-394 470320-350 470320-352 CA470318-690 CA470318-732 CA470318-700 470320-354 CA470318-734 CA470318-702 470320-412 CA470318-698
Amino Labs Canvas Kit™
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Create living paintings with colorful bacteria.
  • Safety First! The kit uses a non-pathogenic strain of E. coli (K-12)
  • Fun first step into learning about genetic engineering and BioArt
  • Great for biology or visual arts class
  • Recommended for use with Amino LabsDNA Playground™ (available separately)
Colored bacteria included are purple, cyan (fluorescent), and magenta (fluorescent). If you have already engineered cells with an Engineer-it Kit and still have them, you can also use those cells with a Canvas Kit™! When you're done creating, lock in your colorful creation with the Keep-It™ Resin kit.
In the Canvas Kit™ students will: Make 'painting canvases', called LB agar petri dishes. Paint or stencil the colorful bacteria included in the kit onto three canvases and incubate them. Like an invisible ink, you won't see them at first, but as they grow and divide they will become visible between 12 to 48 hours and change color. Your painting will emerge!
Additional equipment required for Canvas Kit™ but not included: Microwave or hot-plate, and Amino Labs DNA Playground™ (In the absence of a DNA Playground™, you will also need an incubator and timer.)
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty