Would you like to log in again?
Amino Labs Junior R&D Kits™
Supplier: AMINO LABS NORTH INCORPORATED SE
WWE014-AMP WWE014-SPE_CA WWE014-ERY_CA WWE014-SPE WWE014-CHLOR WWE014-AMP_CA WWE014-CHLORCA WWE014-KAN WWE014-ERY WWE014-KAN_CA
470320-396EA 198.19 USD InStock
CA470318-718 CA470318-714 CA470318-716 470320-396 CA470318-722 470320-398 470320-400 470320-402 470320-404 CA470318-720
Amino Labs Junior R&D Kits™
Educational Materials Biology Educational Materials Genetics Learning Activities
This kit enables a junior researcher to complete DNA transformations with their own engineered DNA and cells.
  • Kit utilizes user-supplied engineered cells for the extraction
  • Great next step after completing the Engineer-It™ or RGB Kit™ (also available)
  • Recommended for use with Amino Labs DNA Playground™ (available separately)
Have you purchased or acquired a DNA plasmid that you'd like to put into E. coli bacteria? Have you completed some ligations and want to transform E. coli with your ligated products? Then these kits are for you! You supply the cells and DNA plasmid, and simply select the kit with the type of antibiotic you need for your experiments, and get ready for transitioning to your own R&D project.
Additional equipment required but not included: Microwave or hot-plate, and Amino Labs DNA Playground™ (In the absence of a DNA Playground™, you will also need an ice bath, water bath, thermometer, timer, and Incubator).
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery information: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty