Would you like to log in again?
Countertop Aquaculture System
Supplier: R&D AQUAFARMS SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2.5CTA CTA HEATER-100
470320-440KT 455.35 USD InStock
470320-440 470320-454 470320-468
Countertop Aquaculture System
Educational Materials Biology Educational Materials Ecology Learning Activities
A compact multi stage system.
  • Available in two formats
  • Easily monitor conditions at each stage
  • Small enough for a classroom
  • Easy to set up and run
This small version of the full size three barrel system fits in an area just two feet square. It is a complete turnkey system requiring only a few connectors screwed together by hand making it very easy to clean or move in a matter of minutes. Made from recycled water carboys makes observing each stage of the system easy. The totally self contained system requires only one 120 V outlet to run the air pump and optional heater. All containers, pipe, connectors, filter material, air pump, and air stone are included.
In the three barrel system water flows between a fish habitat, the growth tank and a filtration tank. Individual tanks makes detailed monitoring easy for students. The growth area supports a one plant hydroponic raft (included). For more growing area, with a similar footprint, the 2.5 barrel replaces the filtration tank with a bisected carboy that sits on top of the fish habitat, and water return. The bisected carboy serves dual purpose as growth chamber and biological filter tank.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty