Would you like to log in again?
Aquaponics Growracks
Supplier: R&D AQUAFARMS SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2BGRA GRA
470320-442KT 1405.9 USD InStock
470320-442 470320-444
Aquaponics Growracks
Educational Materials Biology Educational Materials Ecology Learning Activities
Easily expand your growing output!
  • Available as a stand alone or with barrels
  • Can be used with an existing tank or system
  • Easily assembled
This grow rack can be used with an existing tank or aquarium or be placed on top of optional 55 gallon fish/filter barrels (sold separately). It can also be used next to a pond outdoors. Four trays can support a total of 24 plants. Each tray has adjustable water depths. Higher water levels for starting new sprouts. Lower water levels for use with NFT for established plants. Tray covers can be removed for ease of cleaning.
Dimensions when set up (Rack only): 48W×25D×18" H
Delivery information: Includes: rack, trays, net pots, rock wool cubes, water pump, supply input. Ships flat and snaps together for easy assembly.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty