Would you like to log in again?
Ward's® Incubating Shakers
Supplier: Benchmark Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ward's®
H2012-E_WARD H2012_WARD H1001-M_WARD H3501_WARD H1001-M-E_WARD H3501-E_WARD H2010-E_WARD H2010_WARD
470320-570EA 12354.99 USD InStock
470320-570 470320-580 470320-572 470320-582 470320-574 470320-584 470320-576 470320-578
Ward's® Incubating Shakers
Shakers and Mixers Shaking Incubators
These incubating shakers provide digitally regulated temperature with precise orbital mixing.
  • Great for cell culture or bacterial cell growth
  • Non-slip rubber platform mat included
  • Horizontally circular orbit for aeration and mixing
  • Optional platform available for use with MAGic Clamp™ system
  • Two year warranty
The XL Incu-Shaker is available with or without refrigeration. Mini Incu-Shaker is available with or without CO₂ incubation.
All units are compatible with the optional MAGic Clamp System which allows use of the MAGic Clamp™ attachment line, for quick-changes between flask clamps of different sizes. Additional purchase of MAGic Clamp Platform required. See product resource tab for list of compatible accessories.
Ordering information: Supplied with a non-slip, rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays. Optional platform available for use with MAGic Clamp™ system, this must be ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty