Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Red Laser Pointer

Supplier: United Scientific Supplies
RLP006
470320-708EA 9.6 USD InStock
470320-708
Red Laser Pointer
Laser Pointers
Economical laser pointer, useful for many optical experiments.

  • Max. power 5 mW (Class 3R / IIIa)
  • Approximate wavelength 650 nm
  • Range 100 m (300')
  • Presentation aid, and perfect for diffraction experiments

Economical laser pointers are useful for many optical experiments. Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip. The lasers are Class 3R.

Red laser pointer has a maximum output of 5mW and a wavelength of 650 nm (±10 nm). Range of beam is 100 meters. Expected operating life is 1000 hours.

Ordering information: Operates on two 'AAA' batteries (not included).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty