Would you like to log in again?
Light Leaper Accessory
Supplier: United Scientific Supplies
LLA001
470320-722EA 28.8 USD InStock
470320-722
Light Leaper Accessory
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Vividly illustrate electrical charges.
  • Easy to set up design
  • Works with Wimshurst or Van de Graff generators
This is a popular accessory for electrostatic demonstrations. An insulating printed circuit board carries a metallic conduction path with six breaks and a binding post at each end.
When connected to an operating electrostatic generator, simultaneous sparks occur in each of the breaks as charge flows between the poles of the generator.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty