Would you like to log in again?
Reflection and Refraction Tank
Supplier: United Scientific Supplies
RRT002
470320-750EA 105.8 USD InStock
470320-750
Reflection and Refraction Tank
Educational Materials Physics Educational Materials Optics Learning Activities
Classical demonstration of refraction and internal reflection.
  • Easy to position laser pointer
  • Includes manual
This product effectively demonstrates the laws of reflection and refraction. The 16 cm diameter acrylic water tank has a 360 degree scale and a moveable arm carrying a laser activated by a push-button. Thumb screws on the side of the laser harness can be adjusted to finely position the laser beam.
Ordering information: Requires two AAA batteries (not included). Includes activity guide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty