Would you like to log in again?
Media/Storage Bottles, Square, Clear PET, United Scientific
Supplier: United Scientific Supplies
BMPE500 BMPE1000 BMPE250
470320-956PK 50.7 USD InStock
470320-956 470320-958 470320-954
Media/Storage Bottles, Square, Clear PET, United Scientific
Bottles Plastic Bottles
Clear PET bottles for storage.
  • Supplied with polypropylene cap
  • Sterilized using gamma radiation
  • Individually bagged
Clear, square, disposable polyethylene terephthalate (PET) bottles are space-saving and convenient to handle. Useful for mixing, sampling, and storage applications. Includes a blue, liner-less polypropylene plug-seal autoclavable cap and features raised graduations. Neck int.Ø is 37 mm.
PET is transparent, lightweight, and break-resistant. Bottles can be used in environments ranging from −40...+50 °C. PET cannot be autoclaved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty