Would you like to log in again?
Filtering Bottle
Supplier: United Scientific Supplies
FFB5340-10000
470320-968EA 440.35 USD InStock
470320-968
Filtering Bottle
Flasks Filter Flasks
Thick glass walls and sturdy construction for vacuum filtration.
  • Enable to withstand vacuum filtration without imploding
This 10 L bottle/flask is perfect for filtering large volumes of liquid. Graduated in 1000 ml increments on a scale from 2000 ml to 8000ml with an accuracy of ± 5%. Neck size fits a #13 rubber stopper. The hose barb has an O.D. of ½".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty