Would you like to log in again?
Ward's® Bat Anatomy Museum Mount
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ward's®
WARD470323-066
470323-066EA 624.99 USD InStock
470323-066
Ward's® Bat Anatomy Museum Mount
Specimens Preserved Specimens Display Specimens
Museum mounts are the ideal supplement for classroom dissection.
  • High quality display mount
  • Dissection reference for students
  • Quiz mechanism for teachers
An excellent teaching tool for general mammalian anatomy.
Museum mounts are expertly crafted by our own laboratory technicians to show external and internal anatomy in great detail. Specimens are encased in acrylic with formalin for indefinite preservation.
Museum mounts can be used as a reference for teachers and students or as a quiz mechanism by covering up the key on the back of the case.
Key includes:: 1. Heart 2. Lungs 3. Diaphragm 4. Liver 5. Stomach 6. Spleen 7. Ileum 8. Bladder 9. Anus.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty