Would you like to log in again?
Eisco Light Wall Glass Test Tubes with Beaded Rim
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CH0718GPK24 CH0718AAPK48 CH0718EPK48 CH0718D01PK48 CH0718FPK24 CH0718BPK48 CH0718CPK48 CH0718HPK24 CH0718F01PK24 CH0718APK48
470323-166PK 12.6 USD InStock
470323-166 470323-144 470323-154 470323-142 470323-152 470323-148 470323-158 470323-146 470323-156 470323-150
Eisco Light Wall Glass Test Tubes with Beaded Rim
Tubes Test Tubes
Light wall test tube with beaded rim for general use.
  • Made from high quality Borosilicate 3.3 glass
  • Feature fire polished, beaded rim, and uniform wall thickness
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient- can withstand direct flame, high temperatures and thermal shocks
Eisco Labs Borosilicate glass has the following heat tolerances: strain point –515 °C; annealing point 565 °C; softening point 820 °C. They manufacture their own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty