Would you like to log in again?
InTemp® Multiple Use Dry Ice Logger
Supplier: ONSET COMPUTER CORPORATION SE
CX603
470323-332EA 211.49 USD InStock
470323-332
InTemp® Multiple Use Dry Ice Logger
Data Loggers
The InTemp® CX603 Dry Ice data logger is a Bluetooth® Low Energy (BLE)-enabled multiple-use logger that records temperature data for up to 365 days.
  • IATA battery compliant
  • Three Point NIST Certificate of Calibration, valid for one year from date of purchase
  • Anti-cut sheath
  • Integrates with mobile and cloud platforms for a 21 CFR Part 11 compliant solution
  • Compact and lightweight – ideal for shipping pharmaceuticals
  • No need to open containers to access data
  • Ensures secure data via encrypted technology
The CX603’s built-in 1.5 m probe measures temperatures as low as −95 °C (−139 °F) and includes a protective sheath to prevent accidental cutting of the cable, and a clip for secure mounting.
Using the InTemp® app on your mobile device, you can easily configure the CX603 with one of multiple preset configurations, or set up a custom configuration. The logger has a built-in alarm to alert you to excursions. And logger reports can be quickly downloaded to your phone or tablet and uploaded to an InTempConnect® account, where you can analyze and export data as needed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty