Would you like to log in again?
HOBO MICRORX STATION W/ SOLAR WL
Supplier: ONSET COMPUTER CORPORATION SE
Microrx Water Level Station With Solar^ HOBO* MicroRX Water Level Station with built-in solar panel, Non-vented ceramic water level sensor with a choice of four ranges, Integrated 1.7 W solar panel with rechargeable battery pack, Integrated barometric pressure sensor, For Outdoor environments
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOBO MICRORX STATION W/ SOLAR WL 470323-354 RX2104 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty