Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HOBO® Water Level Data Loggers
Supplier: ONSET COMPUTER CORPORATION SE
U20-001-01 U20-001-04 U20-001-03-TI U20-001-04-TI U20-001-02 U20-001-01-TI U20-001-02-TI U20-001-03
470323-396EA 872.79 USD InStock
470323-396 470323-394 470323-400 470323-398 470323-404 470323-402 470323-408 470323-406
HOBO® Water Level Data Loggers
Data Loggers
The HOBO® water Level data logger features high accuracy at a great price and ease-of-use, with no cumbersome vent tubes or desiccants to maintain.
  • Lightning protection - no long signal wires, and electronics are shielded in stainless steel housing for use in freshwater (see the Water Level logger sensor location drawing)
  • HOBOware Pro software provides easy conversion to accurate water level reading, fully compensated for barometric pressure (see demo) temperature, and water density
  • Multiple-rate sampling (see demo) allows faster sampling at critical times such as when pumping starts or stops
  • Available in 4 depth ranges
  • Ideal for use in wells, streams, lakes, wetlands and tidal areas
  • No-vent-tube design for easy reliable deployment
  • Available in stainless and titanium versions
  • Durable ceramic pressure sensor
  • 3-point NIST-traceable calibration certificate included
This data logger is ideal for recording water levels and temperatures in shallow wells, streams, lakes and freshwater wetlands. For saltwater deployments, such as brackish wetlands and tidal areas, see HOBO® U20 Water Level Titanium.
Ordering information: This logger requires HOBOware Pro software and either a waterproof shuttle or the optic USB base Station for configuration and data offload. For more information on required HOBOware software (not included), contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty