Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HOBO® Water Level 100' Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
U20L-02
470323-412EA 442.89 USD InStock
470323-412
HOBO® Water Level 100' Data Logger
Data Loggers
The HOBO® U20L-02 is a low-cost, research-grade water level data logger for continuously measuring water level and temperature in a wide range of underwater environments.
  • Self-contained non-vented design enables easy deployment
  • Ideal for use in both fresh and saltwater environments, including wells, streams, lakes, wetlands, and tidal areas
  • Depths up to 100 feet
  • Durable ceramic pressure sensor withstands freezing
  • HOBOware® Pro software provides easy conversion to accurate water level reading, fully compensated for barometric pressure, temperature, and water density
It features 0.1% measurement accuracy, a polypropylene housing for use in both fresh and salt water, and a non-vented design for convenient and hassle-free deployment.
Ordering information: This logger requires HOBOware® Pro software and either a U-DTW-1 Waterproof Shuttle or the Base-U-4 Optic USB Base Station for configuration and data offload. For more information on required HOBOware software (not included), contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty