Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HOBO® Pendant® Temperature/Alarm Data Loggers
Supplier: ONSET COMPUTER CORPORATION SE
UA-001-08 UA-001-64
470323-426EA 94.49 USD InStock
470323-426 470323-424
HOBO® Pendant® Temperature/Alarm Data Loggers
Data Loggers
This value-priced HOBO Pendant® temperature/alarm data logger is both miniature and waterproof, making it a sensible monitoring solution for specific temperature monitoring applications.
  • 8K model stores 6500 readings
  • 64K model stores of 52000 readings
  • Low-cost temperature with alarm indication
  • Waterproof housing for wet or underwater use up to 30 m
  • Data readout in less than 30 seconds via fast optic USB interface
This logger has one channel and is recommended for short-term shipping or long-term monitoring. This data logger features a visible LED light indicator, which warns users when preset parameters have been exceeded.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty