Would you like to log in again?
FLEX SMART ANALOG MODULE
Supplier: ONSET COMPUTER CORPORATION SE
Flexsmart Analog Input Module 12-Bit^ HOBO* FlexSmart Analog Input Module, 12-bit, Handles two single-ended input channels with user-configurable ranges of 0 to 20 mADC or 0 to 20 VDC which is suitable for 2.5-, 5-, and 10-volt sensors
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLEX SMART ANALOG MODULE 470323-442 S-FS-CVIA Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty