Would you like to log in again?
SIM Lipoma Surgical Models
Supplier: SIMULATED INANIMATE MODELS LLC SE
860000027001 860000027002
470323-450EA 434.25 USD InStock
470323-450 470323-448
SIM Lipoma Surgical Models
Educational Materials Biology Educational Materials Human Biology Learning Activities
A single use surgical simulator with a real tissue feel.
  • Made of water based hydrogel
  • Imaging compatible
  • Excellent tissue characteristics
SIM prostate phantoms are composed of a water-based hydrogel material that enables superior ultrasound imaging properties versus the competition and are compatible with both standard side-fire and end-fire ultrasound probes. These trainers are ideal for training in Lipoma diagnosis and removal. Each trainer contains a large lipoma for removal, it can be palpated and moved, biopsied, and surgically removed. The advanced trainer also allows for imaging tests prior to removal.
Packaging: For greatest shelf life, refrigerate on arrival, and leave sealed until used.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty