Would you like to log in again?
HOBO® External Temperature/RH Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
MX2302A
470323-472EA 261.19 USD InStock
470323-472
HOBO® External Temperature/RH Data Logger
Data Loggers
The HOBO MX2302 features external temperature and relative humidity sensors on a 2 m cable.
  • Convenient wireless setup and download via Bluetooth Low Energy (100-foot range)
  • Record a maximum 84650 measurments
  • Visual alarms alert you to out-of-range conditions
  • Compact, weatherproof housing with built-in mounting
  • View current data in real time with Onset’s free HOBOmobile app
  • Share downloaded data easily from your mobile device
Leveraging Bluetooth Low Energy, the logger enables easy, fast set up and data download directly from your mobile device and provides high-accuracy measurements in harsh outdoor environments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty