Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HOBO® 2× External Temperature Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
U23-003
470323-514EA 287.89 USD InStock
470323-514
HOBO® 2× External Temperature Data Logger
Data Loggers
A weatherproof data logger with two external soil/water temperature probes on 6' cables for fast sensor response and deployment in tight spaces.
  • Weatherproof housing for use in outdoor or condensing environments
  • High accuracy
  • Small-diameter external sensor versions for measurements in tight spaces
  • Optic USB interface for fast and reliable download
Compatible with HOBOware® and HOBOware® Pro software for logger setup, graphing, and analysis.
Ordering information: Requires HOBOware® software and a USB cable. (NOTE: A USB cable is included with HOBOware® Pro CD or USB software purchase). HOBOware® Pro is required when using the U-DTW-1 waterproof shuttle.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty