Would you like to log in again?
HOBO® Temperature and Relative Humidity Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
UX100-003
470323-522EA 143.39 USD InStock
470323-522
HOBO® Temperature and Relative Humidity Data Logger
Data Loggers
The Temperature/Relative Humidity data logger records temperature and relative humidity (within 3.5% accuracy) in indoor environments with its integrated sensors.
  • Easy-to view LCD display
  • Large memory capacity
  • Visual high and low alarm thresholds
  • New Burst and Statistics logging modes
  • User-replaceable relative humidity sensor
  • Compatible with HOBOware and HOBOware Pro software for logger setup, graphing and analysis
The data logger is ideal for monitoring occupant comfort in office buildings, tracking food storage conditions in warehouses, logging temperature trends in server rooms, and measuring humidity levels in museums.
Ordering information: HOBO data loggers require HOBOware software and a USB cable. (Note: A USB cable is included with HOBOware Pro CD).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty