Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HOBO® Plug Load Data Logger, Onset
Supplier: Onset
UX120-018
470323-538EA 402.3 USD InStock
470323-538
HOBO® Plug Load Data Logger, Onset
Data Loggers
A high-accuracy, easy-to-deploy data logger that measures and records the power and energy consumption of 120 V plug loads from office equipment, vending machines, and other devices.
  • 0.5% measurement accuracy enables more reliable plug load energy data
  • 'Meter Mode' provides instant viewing of real-time plug load variables
  • Provides time-stamped and totalized load data for detailed energy analysis
  • Large memory enables faster logging intervals for more comprehensive plug load profiles
This UL-certified logger provides 0.5% measurement accuracy with a measurement resolution of 1 watt, making it an ideal choice for building audits where detailed energy use data is required. The rugged housing is designed to withstand the rigors of real-world handling, providing reliable performance despite bumps, drops, and use under desks.
Ordering information: Requires HOBOware software and a USB cable.(Note: A USB cable is included w/HOBOware Pro CD or USB software purchase).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty