Would you like to log in again?
HOBONET SOLAR RADIATION SENSOR
Supplier: ONSET COMPUTER CORPORATION SE
Sensor Hobonet Solar Radiation 900 Mhz^ HOBOnet Solar Radiation (Silicon Pyranometer) Sensor, Measurement range of 0 to 1280 W/m2 over a spectral range of 300 to 1100 nm, Enclosed in an anodized aluminum housing with acrylic diffuser and O-ring seal, 900 MHz wireless mesh self-healing technology
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOBONET SOLAR RADIATION SENSOR 470323-646 RXW-LIB-900 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty