Would you like to log in again?
HOBOnet Wireless Temperature Sensor
Supplier: ONSET COMPUTER CORPORATION SE
RXW-TMB-900
470323-660EA 310.05 USD InStock
470323-660
HOBOnet Wireless Temperature Sensor
Data Loggers
The HOBOnet Wireless Temperature Sensor provides a cost-effective and scalable solution for web-enabled monitoring of air, soil, and water temperatures.
  • Stainless–steel sensor tip
  • Sensor and cable rated for immersion in water or soil up to 50 °C for up to 1 year
  • 450 to 600 meter (1500 to 2000 feet) wireless range and up to five hops
  • Up to 50 wireless sensors or 336 data channels per HOBO RX station
  • Simple button–push to join the HOBOnet wireless network
  • Onboard memory to ensure no data loss
  • Powered by rechargeable AA batteries and built–in solar panel
Ordering information: A complete system requires at least one HOBO RX3000 Remote Monitoring Station, a HOBOnet Wireless Manager, and a HOBOnet Wireless Sensor, OR one HOBO MicroRX Station (which has an integrated HOBOnet Wireless Manager) and a HOBOnet Wireless Sensor. HOBOnet Wireless Repeaters can be added to extend the range.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty