Would you like to log in again?
WIND SPEED SENSOR WITH 3M CABLE
Supplier: ONSET COMPUTER CORPORATION SE
Smart Sensor Wind Speed 0 To 170 Mph^ Wind Speed Smart Sensor, Measure wind speed from 0 to 76 m/s (0 to 170 mph), This smart sensor provides data reporting average wind speed and highest 3 second gust for each logging interval, The starting threshold is 1 m/s (2.2 mph), with 3m cable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIND SPEED SENSOR WITH 3M CABLE 470323-738 S-WSB-M003 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty