Would you like to log in again?
Cloud Formation Apparatus
Supplier: GSC INTERNATIONAL SE
4-30101
470323-886EA 20.65 USD InStock
470323-886
Cloud Formation Apparatus
Educational Materials Earth Science Educational Materials Meteorology Learning Activities
Demonstrate the principles of cloud formation, even when there isn't a cloud in the sky, with the cloud formation apparatus.
When atmospheric pressure is lowered in the chamber and smoke particles are introduced, a nucleus for the formation of water droplets is produced. When the bulb is compressed and then suddenly released, a cloud is formed inside the flask.
Delivery information: Instructions included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty