Would you like to log in again?
mBot Ranger Robot Kit
Supplier: THE DOUGLAS STEWART CO.,INC
121867
470324-126EA 232.69 USD InStock
470324-126
mBot Ranger Robot Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
This robot kit is a favorite in the classroom.
  • Chassis: Aluminum alloy
  • Processor: ATmega2560
  • Coding language: Block-based or Arduino
  • Communication mode: USB or Bluetooth
The mBot Ranger is an intermediate robotics kit designed for STEAM educators and robotics enthusiasts. Enjoy versatile hands-on activities in class, in workshops, at home, and more! It has three configurations that help children explore the mechanical engineering and design: robot tank, self-balancing robot, and racing car. Conquer all terrains like flat roads, grassland, gravel, and sandy beach.
When paired with the free programming apps and instructional resources, it provides a visual programming experience for all users. Supports Scratch and Arduino C programming, so kids can learn coding from beginner to expert.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty