Would you like to log in again?
Aquatic Insects Riker Mount R288
Supplier: WHITE OWL ENTERPRISES SE
R288
470324-272EA 62.15 USD InStock
470324-272
Aquatic Insects Riker Mount R288
Specimens Preserved Specimens Display Specimens
Riker mount of aquatic insects.
  • High quality display mount
Use this display to bring the world of aquatic insects into the classroom, or to identify common, aquatic insect life that you observe in the field. This 6×8" exhibit is fully labeled and features the aquatic stages of 10 common insects.
Typically featured are the damselfly naiad, dragonfly naiad, caddisfly larva, mosquito larva, phantom gnat larva, water strider, backswimmer, water boatman, water scavenger beetle and crawling water beetle.
Ordering information: Specimens may vary with availability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty