Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HOBO® Analog/Temperature/RH/Light Data Logger
Supplier: ONSET COMPUTER CORPORATION SE
SD-VOLT-24 T-ASH-G2-100 SD-TEMP-06 SD-VOLT-2.5 T-ASH-G2-500 SD-TEMP-01 T-CON-ACT-150 T-DCI-F300-XXX T-ASH-G2-200 T-CDI-5200-10S SD-MA-420 T-CDI-5400-20S SD-VOLT-10 MX1104 SD-TEMP-20 SD-VOLT-05 T-VER-PX3UL T-CON-ACT-300 SD-TEMP-50
470324-608EA 240.69 USD InStock
470324-608 470324-618 470335-926 470335-928 470335-922 470335-944 470335-934 470335-946 470335-924 470324-602 470335-936 470335-940 470324-604 470335-930 470324-606 470335-920 470335-942 470335-932 470330-995
HOBO® Analog/Temperature/RH/Light Data Logger
Data Loggers
This data logger measures and transmits temperature, relative humidity and light intensity data wirelessly.
  • Bluetooth communication for wireless data offload
  • 16-bit resolution for highly-accurate measurements
  • Stores 1.9 million measurements for longer deployments between offloads
  • New self-describing sensors with automatic configuration for fast deployment
  • Compatible with existing sensors for a wide range of indoor monitoring
  • Audible and visual LCD-screen alarms notify you if sensor becomes unplugged or a reading exceeds set thresholds
Includes an external analog input to attach a variety of additional sensors.
This self-contained, bluetooth-enabled, wireless data logger lets you use your mobile device or Windows computer and Onset's free HOBOconnect app to access data, at any time, when within 100' of the logger. Configure the logger, read out data, view data in graphs, check the operational status of loggers, configure alarm notifications, and share data files - with no dedicated equipment beyond a mobile device or laptop.
Ordering information: Requires HOBOconnect® app and compatible mobile device or Windows computer. Add the MX gateway for remote access to your data in our cloud-based HOBOlink® software. For more information on required HOBOware® software (not included), contact your Avantor representative.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty