Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
J&D Sensing EG4100 Series Current Transformer Sensor
Supplier: ONSET COMPUTER CORPORATION SE
T-EG-0940-0200 T-EG-0390-0050 T-EG-0390-0075 T-EG-0630-0050 T-EG-1420-0400 T-EG-1420-0600 T-EG-0940-0100 T-EG-0630-0100
470324-640EA 58.49 USD InStock
470324-640 470324-650 470324-642 470324-644 470324-646 470324-648 470324-636 470324-638
J&D Sensing EG4100 Series Current Transformer Sensor
Data Loggers
Split-core CTs offer a well constructed hinged body and are accurate to ±1% from 1 to 100% of their stated current rating.
  • Multiple current ratings
  • Small size, light weight
  • Simple installation
  • Overvoltage protection circuit
  • Pin-hinged at pivot point
The small interval for physical size options 10 mm (0.39"), 24 mm (0.94"), and 36 mm (1.42") allow for a flexible installation.
Ordering information: Requires HOBOware software and a USB cable. Compatible with H22-001,U30-NRC,UX90-001M,RX3000, U30-NRC-SYS-C, and UX120-017M.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty