Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco® Borosilicate Glass Narrow Neck Erlenmeyer Flask, 100 ml

Supplier: EISCO SCIENTIFIC LLC
CH0424D
470324-792EA 3.9 USD InStock
470324-792
Eisco® Borosilicate Glass Narrow Neck Erlenmeyer Flask, 100 ml
Flasks Erlenmeyer Flasks
High-quality, graduated 100 ml Erlenmeyer flask with a narrow neck.

  • Glass Erlenmeyer (conical) flask
  • Made with high-quality Borosilicate 3.3 glass
  • Features 25 ml white enamel graduations

Eisco® Borosilicate glassware has the following heat tolerances:
Strain point: −515°C
Annealing point: 565 °C
Softening point: 820 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty