Would you like to log in again?
Eisco Economy Microscope Slide Viewer
Supplier: EISCO SCIENTIFIC LLC
BI0276BV
470324-796EA 22.75 USD InStock
470324-796
Eisco Economy Microscope Slide Viewer
Microscopes
A fun introduction to the world of microscopy for students.
  • Lightweight and easy to use
  • Uses ambient light
  • Wireless, no battereies or light bulbs required
  • Compact design for easy storage and handling
This economical microscope slide viewer is light weight and economical. The unit does not require batteries or to be plugged into to an electrical source. Light from a lamp or natural light can be directed to the white surface and the light will reflect into the viewer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty