Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Flip-Top Hinged Cardboard Freezer Boxes
Supplier: MTC BIO, INC.
R2700-O_WARD R2781-P_WARD R2700-P_WARD R2781-A R2781-R R2700-P R2700_WARD R2700-B_WARD R2700-A R2700-R R2781-A_WARD R2700 R2781-O R2700-G R2781-O_WARD R2700-B R2700-R_WARD R2700-O R2781_WARD R2781-R_WARD R2781-G R2700-G_WARD R2781-P R2781-B_WARD R2700-A_WARD R2781-G_WARD R2781-B R2781
470324-926PK 23.55 USD InStock
470324-926 470324-948 470324-928 470324-984 470324-940 470324-942 470324-986 470324-988 470324-944 470324-968 470324-946 470324-980 470324-982 470324-938 470324-930 470324-974 470324-952 470324-976 470324-932 470324-934 470324-978 470324-936 470324-990 470324-970 470324-992 470324-950 470324-994 470324-972
Flip-Top Hinged Cardboard Freezer Boxes
Boxes Cryoboxes
Our hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating.
  • Accommodates 1.5 and 2.0 ml tubes, as well as most 2.0 ml cryogenic vials
  • Alpha-numeric coding helps sample identification
  • Standard sized boxes will fit in your existing racks
  • Available in six colors, in two different tube configurations
Choose between 81 or 100-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, and so on).
Ordering information: Available in white and five individual colors, plus a 'Rainbow Pack'. Boxes are shipped fully assembled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty