Would you like to log in again?
VictoryXR® AR Victor Torso
Supplier: VICTORY VR INC SE
2DTORSO VICTORMODEL1
470325-086EA 300.75 USD InStock
470325-086 470325-064
VictoryXR® AR Victor Torso
Educational Materials Biology Educational Materials Physiology Learning Activities
Take the next step in anatomical studies.
  • Augmented reality organs
  • Audio information embedded in the app
  • Free mobile app
Victor the torso is a new approach to learning about body systems and organs. Using the free augmented reality app, students can select, and examine individual organs at their own pace. A double click provides information about the organ, including landmarks and structural information. Each set of organs is set with a different chest plate, so you can keep a lecture focused on a specific region.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty