Would you like to log in again?
Shark Week Kits
Supplier: GENOSENSOR CORPORATION
7003 7004
470325-174EA 156.5 USD InStock
470325-174 470325-172
Shark Week Kits
Educational Classroom Kits and Activities
Teach fundamental concepts of genetic diversity and real world applications in marine biology.
Marine biologist have identified a new species of shark that washed up on the beach. In an attempt to learn as much about the shark as they can, they have decided to analyze the sharks stomach contents to identify its main food source.
Plasmid DNA is used to emulate the genetic diversity seen in biology. Four plasmid samples are included in the kit, one representing DNA from the main food source, and three for comparison against the unknown food DNA.
Kit I: Allows students to perform a restriction digest of four DNA samples and compare their sizes via agarose gel electrophoresis.
Ordering information: Equipment needed but not included: Heat block/water bath for restriction. Thermal cycler for PCR, microcentrifuge, vortex for sample preparation. Electrophoresis equipment/reagents including transilluminator/light box for analysis.
Delivery information: Kit includes perishable materials, please provide a delivery date at check-out. Kits contain reagents and instructions to download procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty