Would you like to log in again?
Insect Metamorphosis Comparison
Supplier: WHITE OWL ENTERPRISES SE
R277
470325-206EA 81.55 USD InStock
470325-206
Insect Metamorphosis Comparison
Specimens Preserved Specimens Display Specimens
Riker mount of insect metamorphosis for comparison.
  • High quality display mount
Compare the stages of simple (hemimetabolous) metamorphosis to those of complete (holometabolous) metamorphosis with this display.
In simple metamorphosis the insect has only 3 life stages: egg, nymph (looks like a smaller, wingless adult), and adult. This type of development is shown using specimens from a grasshopper. With complete metamorphosis there are 4 distinct stages: egg, larva (a worm-like stage), pupa (non-feeding and sessile), and finally adult. These stages are represented using butterfly specimens.
All specimens are clearly labeled and presented in a 6×8" display.
Ordering information: Exact species will vary with availability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty