Would you like to log in again?
Hermle Z216MK High-Speed, Refrigerated Microcentrifuges
Supplier: Ward's
Z216-MK Z216-K-CMB Z216-2420 Z216-3202 Z216-6420 Z216-2420S Z216-3020H Z216-K-BND44 Z216-2420H Z216-24HEM Z216-4420 Z216-1205
470230-626EA 7790 USD InStock
470230-626 470230-636 470313-070 470230-634 470230-632 470325-214 470230-642 470230-630 470325-212 470230-640 470230-628 470230-638
Hermle Z216MK High-Speed, Refrigerated Microcentrifuges
Centrifuges Benchtop Centrifuges
Compact, quiet, and powerful.
  • Bundles available with our most popular rotors
  • Automatic lid lock prevents opening while rotor is in motion
  • Imbalance detection system powers down motor when improperly loaded
  • Brushless induction drive and microprocessor control
Purchase a bundle and save: Z216MK is available pre-bundled with one of the two most popular rotors. Or if you need something more specialized, purchase the centrifuge alone and select from several available rotor options.
The Z216MK has a compact design and quiet operation; refrigeration keeps samples at temperatures as low as 4 °C during a continuous run at 20000×g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty