Would you like to log in again?
Aluminum Dissecting Trays with Wax
Supplier: United Scientific Supplies
DSPA02-W DSPA01-W
470339-910EA 59.95 USD InStock
470339-910 470325-656
Aluminum Dissecting Trays with Wax
Trays/Dishes/Pots Dissecting Trays
Made of aluminum and filled with a layer of paraffin wax, these dissecting pans will give you years of service with minimal maintenance.
  • Easy to Clean
  • Ideal Size for Smaller Specimens
  • Easy to Store
The paraffin holds T-pins securely without breaking down with repeated use. The wax resists absorption of most solutions allowing for a simple rinse and pour clean-up. This size of tray is ideal for smaller dissections like Clams, Squid, earthworm, locust, and most perch or smaller frogs.
Refill wax is available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty