Would you like to log in again?
Flask Form Electroscope, United Scientific
Supplier: United Scientific Supplies
ELFL01
470325-664EA 13.85 USD InStock
470325-664
Flask Form Electroscope, United Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Classroom electroscope housed in a 250 ml borosilicate flask.
  • Activity guide included
This electroscope is housed in a 250 ml borosilicate flask with a metal rod attached to a rubber stopper. A pair of aluminum leaves is suspended from an alligator clip. The knob on top of the rubber stopper accepts a charge. When the charge enters the flask, the aluminum leaves separate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty