Would you like to log in again?
LaMotte® TesTab® Nitrate Test Kit
Supplier: LaMotte
0898 5891
470308-454EA 4.55 USD InStock
470308-454 470326-580
LaMotte® TesTab® Nitrate Test Kit
Environmental Test Kits
Refill for The Comprehensive Water Monitoring Kit 470004-240 and LaMotte® TesTab® Water Investigation Kit 470007-270
  • 100 Tests
  • Range 0, 5, 20, 40 ppm (NH₃)
Each kit contains easy-to-read diagrammed instructions, a color chart, a test factor flash card and all the necessary apparatus and non-hazardous TesTabs® to perform 100 tests
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty