Would you like to log in again?
LaMotte® Lead Screening Test
Supplier: LaMotte
5-0140
470326-588EA 136.99 USD InStock
470326-588
LaMotte® Lead Screening Test
Test Paper Indicator Paper
In only 10 minutes the analyst can detect lead levels above 15 ppb- the EPA action level.
  • Includes 10 tests that come individually sealed with test vial and dropper
  • Results in 10 minutes
  • Detects lead to EPA action level of 15 ppb
Simply immerse the strip in a water sample and read after a 10 minute waiting period.
The multi-test package includes 10 tests and diagrammed instuctions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty