Would you like to log in again?
Cobalt Chloride Paper
Supplier: Precision Laboratories
250-12-8X10
470326-694PK 32.1 USD InStock
470326-694
Cobalt Chloride Paper
Test Paper Indicator Paper
Use this test paper to detect the presence of water or to perform qualitative tests of relative humidity. Size of each sheet: 20 x 25cm. Package of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty