Would you like to log in again?
Elevate Healthcare® Patient Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Elevate Healthcare®
ARES-300M TRN-ARE01 WAR-ARE01 ARES-100 TRN-125 ARES-300D WAR?ARE03 TRN-ARE02 ARES-200 ARES-300 WAR-ARE02 ARES-200D ARES-200M
470327-180EA 395.99 USD InStock
470327-180 470327-184 470338-782 470327-182 470338-780 470327-188 470327-012 470327-186 470327-010 470338-778 470327-178 470327-008 470338-772
Elevate Healthcare® Patient Trainer
Models Human Biology Models
An Emergency Care Manikin you can rely on.
  • Wireless and teatherless control
  • Includes Maestro
Ares is Elevate Healthcare's newest mid-fidelity manikin, designed to fulfill the requirements for Advanced Life Support (ALS), Advanced Cardiac Life Support (ACLS), and emergency care team training within academic programs, hospitals, and emergency care services. Ares is portable, lightweight, and durable, with the right mix of features for life-saving simulation training. Backed by a dedicated customer service team and Academy of expert educators, Ares embodies the exceptional quality and support you expect from Elevate.
With three different options to choose from, there's an option to fit your needs now and for the future. Start with the basic model and upgrade later or get the advanced model for the greatest flexibility. Every version comes with the same basic materials and the Maestro software package with six pre-built simulated clinical experiences (SCE), the differences include the replacement of Tristate eyes with Symeyes (included with advanced and complete) and a Symdefib controller (complete only). Each simulator also includes the first year value assurance policy. Additional, or upgraded, policies can be purchased separately.
Accessories information: For functionality details, and a comparison of what is included in each package please see the additional documents attached to the web page.
Ordering information: These items cannot be returned or refunded. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty