Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blue Phantom® Amniocentesis Ultrasound Trainer

Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BP1610 BP1612
470327-312EA 3659.39 USD InStock
470327-312 470327-310
Blue Phantom® Amniocentesis Ultrasound Trainer
Models Human Biology Models
Adjustable fluid volumes to meet changing training needs.

  • Externally accurate fetal anatomy allowing for 3-D ultrasound training
  • Superb imaging characteristics
  • Self-healing tissue
  • Ultra-durable design
  • Contains anatomical landmarks
  • Practice using ultrasound system controls

Blue Phantom® ultrasound guided amniocentesis training model contains realistic pelvic anatomy including a gravid uterus with a fetus of 18 week gestational age, umbilical cord with both fetal and placental cord insertions, anterior/anterolateral placenta, cervix, a variety of fluid pockets enabling users to begin training utilizing larger fluid pockets and work to develop their skills allowing them to guide needles to smaller pockets of amniotic fluid.

Blue Phantom® expends tremendous effort to match the imaging characteristics of real human tissue, allowing teaching and training under optimal conditions. The best simulation training scenarios are requisite on users experiencing the same procedural realism as performing the procedure on patients. All of our simulation materials match the acoustic characteristics of real human tissue so when you use your ultrasound system on our training models, you experience the same quality you expect from imaging patients in a clinical environment.

Gaining proficiency in using ultrasound requires practice. The self-healing tissue will allow you to repeatedly perform ultrasound guided amniocentesis procedures without giving second thought to wearing out the tissue or calculating the cost per use. The amniocentesis ultrasound training mannequin is constructed using Blue Phantom®’s patented ultra-durable tissue and is extremely realistic in ultrasound imaging characteristics and feels like real human tissue. This self-healing tissue will withstand tremendous use and will save money by dramatically reducing the necessity for purchasing replacement parts. Expect thousands of uses without replacing parts. Fluid removed during ultrasound guided amniocentesis training is easy to refill using a quick fill port.

This medical ultrasound amniocentesis training model will perform well using any ultrasound imaging system configured with the appropriate transabdominal ultrasound transducer. Excellent for specialties including obstetrics and gynecology (Ob/gyn), ultrasound training programs, simulation centers, surgical skills centers, medical education facilities, and ultrasound manufacturers for ultrasound education and demonstrations.

Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty