Would you like to log in again?
Blue Phantom® Ultrasound Training Block Models
Supplier: CAE HEALTHCARE, INC. SE
Blue Phantom®
BPAB210 BPFB140 BPBF200-C BPBV110 BPT195 BPP120 BPTM130 BPO100 BPNB150 BPBF200-G
470327-410EA 838.39 USD InStock
470327-410 470327-416 470327-426 470327-316 470327-314 470327-308 470327-318 470327-320 470327-440 470327-372
Blue Phantom® Ultrasound Training Block Models
Models Human Biology Models
Compact and convenient.
  • Extremely realistic imaging characteristics
  • Constructed of ultra-durable simulated human tissue, for a realistic training experience
  • Feels and cannulates like real human tissue
  • Self-healing tissue
  • Fluid can be injected into the model to verify needle tip location (automatically expelled)
Blue Phantom Select series vascular access ultrasound training models are an excellent training phantom to help clinicians develop, practice, and maintain the skills necessary for ultrasound guided venous access and arterial access procedures. These lightweight models are easily portable for travel and training where you want, when you want. Excellent for clinicians beginning to use ultrasound as well as more advanced users seeking more challenging vascular access insertion technique training. The branched vessels offer multiple training opportunities; independent linear vessels for initial training in addition to multiple overlapping branched vessels for more advanced ultrasound training.
Convex surface contour offers a scanning environment similar to human body habitus. Learn to acquire and interpret imaging. Use any ultrasound imaging system
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty